ContactUsLinkComponent
Human Nucleolin, Mouse IgG2a Fc Tag (NUL-H5254) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Glu 710 (Accessio n # P19338-1). 1 * 100 µG
Catalog # ACRBNUL-H5254100UG
Supplier:  ACRO Biosystems
Human Nucleolin, Mouse IgG2a Fc Tag (NUL-H5254) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Glu 710 (Accessio n # P19338-1). 1 * 100 µG
Catalog # ACRBNUL-H5254100UG
Supplier:  ACRO Biosystems
Frequently Bought Together