ContactUsLinkComponent
Biotinylated Human CD5, Fc,Avitag (CD5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1). 1 * 200 µG
  ACRBCD5-H82F3200UG
Biotinylated Human CD5, Fc,Avitag (CD5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1). 1 * 200 µG
  ACRBCD5-H82F3200UG
Frequently Bought Together