ContactUsLinkComponent
Cynomolgus IL-1 beta, His Tag (ILB-C51H3) is expressed from E.coli cells. It contains AA Ala 117 - Ser 268 (Accession # P79182-1). 1 * 1 mg
Catalog # ACRBILB-C51H3-1MG
Supplier:  ACRO Biosystems
Cynomolgus IL-1 beta, His Tag (ILB-C51H3) is expressed from E.coli cells. It contains AA Ala 117 - Ser 268 (Accession # P79182-1). 1 * 1 mg
Catalog # ACRBILB-C51H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together