ContactUsLinkComponent
Human CLEC4C, Fc Tag (CLC-H5254) is expressed from human 293 cells (HEK293). It contains AA Asn 45 - Ile 213 (Accession # Q8WTT0-1). 1 * 100 µG
  ACRBCLC-H5254100UG
Human CLEC4C, Fc Tag (CLC-H5254) is expressed from human 293 cells (HEK293). It contains AA Asn 45 - Ile 213 (Accession # Q8WTT0-1). 1 * 100 µG
  ACRBCLC-H5254100UG
Frequently Bought Together