ICP METAL STANDARD 31 COMPONENT 1 * 100 mL
ACSDRM057050L1-100
: CPACHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Humidity standards
: ROTRONIC INSTRUMENTS
Buffer solutions in sachets
: Mettler - Toledo
pH buffers, ready to use
: WTW
VWR® Buffer Solutions, Secondary Standards
: VWR Chemicals
VWR®, Flow Cup viscosity standards
: VWR Chemicals