Biotinylated SARS-CoV-2 Spike RBD (L452R, T478K), His,Avitag (SPD-C82Ed) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession QHD43416.1 (L452R, T478K)). The mutations (L452R, T478K) were identified in the SARS-CoV 1 * 25 µG
ACRBSPD-C82ED-25UG
Frequently Bought Together