ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (SPN-C82Ec) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession QHD43416.1). The mutations (T19R, G142D, EF156-157del, R158G, L452R, T478K, D614G, P681R, D950N) we n 1 * 200 µG
Catalog # ACRBSPN-C82EC200UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (SPN-C82Ec) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession QHD43416.1). The mutations (T19R, G142D, EF156-157del, R158G, L452R, T478K, D614G, P681R, D950N) we n 1 * 200 µG
Catalog # ACRBSPN-C82EC200UG
Supplier:  ACRO Biosystems
Frequently Bought Together