Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cleaner and Disinfectant for non-invasive medical devices and nonporous surfaces 1 * 6 Bottle
Catalog # DIVY100941433
Supplier:
Diversey
Expand image
Cleaner and Disinfectant for non-invasive medical devices and nonporous surfaces 1 * 6 Bottle
Catalog # DIVY100941433
Supplier:
Diversey
Frequently Bought Together
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99.8% (GC, on anhydrous basis), PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Isobutanol, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide 99.8+%, extra pure
Supplier: Thermo Fisher Scientific