ContactUsLinkComponent
Sterile sampling bag - w x h: 38 x 56 cm / 15 x 22 pouces - Thickness: 110 µm / 4 mil - Capacity: 13500 mL / 450 oz - Number of stickers: 2 1 * 250 items
Catalog # INSC145385
Supplier:  INTERSCIENCE
undefined
Sterile sampling bag - w x h: 38 x 56 cm / 15 x 22 pouces - Thickness: 110 µm / 4 mil - Capacity: 13500 mL / 450 oz - Number of stickers: 2 1 * 250 items
Catalog # INSC145385
Supplier:  INTERSCIENCE