Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Sterile sampling bag - w x h: 14 x 23 cm / 5.5 x 9 pouces - Thickness: 70 µm / 3 mil - Capacity: 650 mL / 22 oz - Number of stickers: 1 1 * 500 items
INSC145113
:
INTERSCIENCE
Sterile sampling bag - w x h: 14 x 23 cm / 5.5 x 9 pouces - Thickness: 70 µm / 3 mil - Capacity: 650 mL / 22 oz - Number of stickers: 1 1 * 500 items
INSC145113
:
INTERSCIENCE
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals