analogue weather station out of stainless steel with barometer, thermometer and hygrometer weather proof, with attachment for hanging, dimensions 96 x 35 x 282mm, Made in Germany 1 * 1 items
Catalog # HECH43662010
Supplier: GLASWARENFABRIK KARL HECHT
Frequently Bought Together