Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
4 945 201 Centrifuge tubes, nominal volume 10 ml, with round bottom, ungraduated, heavy wall, with screw cap, o.d. 16 mm, length 98 mm 1 * 1 items
Catalog # WITG4.945.201
Supplier:
witeg Labortechnik
Expand image
4 945 201 Centrifuge tubes, nominal volume 10 ml, with round bottom, ungraduated, heavy wall, with screw cap, o.d. 16 mm, length 98 mm 1 * 1 items
Catalog # WITG4.945.201
Supplier:
witeg Labortechnik
Frequently Bought Together
VWR®, Beakers, Low Form, Heavy-Duty
Supplier: VWR Collection
VWR®, Beakers, Tall Form
Supplier: VWR Collection
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR®, Pasteur Pipettes
Supplier: VWR Collection
VWR®, Funnels with Short Stem
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
4 945 202 Centrifuge tubes, 25 ml, with round bottom, ungraduated, heavy wall, with screw cap, o.d. 24 mm, length 98 mm, nominal volume 20 ml 1 * 1 items
Supplier: witeg Labortechnik