Orders LinkContactUsLinkComponent
Polypropylene Muff Covers with Elastic
Polypropylene Muff Covers with Elastic
Polypropylene muff cover, 18" (457 mm), fits 10 - 14" (254 - 356 mm) Ø, White
Catalog # 129-3075
Polypropylene Muff Covers with Elastic
Catalog # 129-3075
Polypropylene Muff Covers with Elastic
Polypropylene muff cover, 18" (457 mm), fits 10 - 14" (254 - 356 mm) Ø, White
Price
2 740,00 €
Log in for availability
1 * 500 items
Product Details & Documents
Keeps cleaned non sterile production hoses protected from external sources of contamination.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Tear and puncture resistant
  • Conveniently packaged in 100 per sealed poly bag
These polypropylene muffs are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
Poly bagged in packs of 100 to help ensure cleanliness up until point of use.
Delivery information: Certificates of Conformance sent with each shipment.
Specifications
  • Colour:
    White
  • Description:
    Polypropylene muff cover, 18" (457 mm), fits 10 - 14" (254 - 356 mm) Ø
  • Seq:
    9