Orders LinkContactUsLinkComponent
Polypropylene Muff Covers with Elastic
Polypropylene Muff Covers with Elastic
Polypropylene muff cover, 12" (305 mm), fits 5,5 - 8" (140 - 203 mm) Ø, White
Catalog # 129-3073
Polypropylene Muff Covers with Elastic
Catalog # 129-3073
Polypropylene Muff Covers with Elastic
Polypropylene muff cover, 12" (305 mm), fits 5,5 - 8" (140 - 203 mm) Ø, White
Price
3 900,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Keeps cleaned non sterile production hoses protected from external sources of contamination.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Tear and puncture resistant
  • Conveniently packaged in 100 per sealed poly bag
These polypropylene muffs are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
Poly bagged in packs of 100 to help ensure cleanliness up until point of use.
Delivery information: Certificates of Conformance sent with each shipment.
Specifications
  • Colour:
    White
  • Description:
    Polypropylene muff cover, 12" (305 mm), fits 5,5 - 8" (140 - 203 mm) Ø
  • Seq:
    7