Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Chlorophenol 99%

2-Chlorophenol 99%

Bottle, 5 g
Catalog # APOSOR52255-5G
Irritant
Environment
Supplier:   Apollo Scientific
2-Chlorophenol 99%
Catalog # APOSOR52255-5G
Supplier:  Apollo Scientific
CAS Number:  95-57-8
2-Chlorophenol 99%

Bottle, 5 g

CAS Number:  95-57-8
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents

Has been used as a reagent for the protection of OH-groups in oligonucleotide synthesis. A study of various cleavage methods showed that the tetramethylguanidine salts of either 4-nitrobenzaldoxime or pyridine-2-carboxaldoxime gave the best results: Tetrahedron Lett., 2727 (1978).

Synonyms:  1-Chloro-2-hydroxybenzene
  • CAS Number:95-57-8
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002159
  • Molecular Weight:128.56 g/mol
  • Boiling Pt:175…176 °C
  • Melting Pt:7…8 °C
  • Melting Pt:7-8 °C
  • Density:1.26 g/cm³ (25 °C)
  • Flash Pt:64 °C
  • Storage Temperature:Ambient
  • EINECS:202-433-2
  • UN:2021
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.