Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Chlorocyclohexanone 95%

2-Chlorocyclohexanone 95%

Bottle, 25 g
Catalog # APOSOR918717-25G
Supplier:   Apollo Scientific
2-Chlorocyclohexanone 95%
Catalog # APOSOR918717-25G
Supplier:  Apollo Scientific
CAS Number:  822-87-7
2-Chlorocyclohexanone 95%

Bottle, 25 g

CAS Number:  822-87-7
Price
30,40 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:822-87-7
  • Formula:C₆H₉ClO
  • MDL Number:MFCD00001626
  • Molecular Weight:132.59 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.