Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Reagecon pH 7.413 DIN 19266 Buffer Solution at 25C 1 * 500 mL
Catalog # REGE107413
Supplier:
REAGECON
Expand image
Reagecon pH 7.413 DIN 19266 Buffer Solution at 25C 1 * 500 mL
Catalog # REGE107413
Supplier:
REAGECON
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals