ContactUsLinkComponent
Analytical Balance ME104TE/00 Maximum Capacity: 120 g Readability: 0.1 mg Minimum Weight (USP, 0.1%, typical): 160 mg Adjustment: External Weighing Pan Diameter: 90 mm Display: 4.5 colour TFT touch screen 1 * 1 items
Catalog # METBME104TE/00
Supplier:  Mettler - Toledo
Analytical Balance ME104TE/00 Maximum Capacity: 120 g Readability: 0.1 mg Minimum Weight (USP, 0.1%, typical): 160 mg Adjustment: External Weighing Pan Diameter: 90 mm Display: 4.5 colour TFT touch screen 1 * 1 items
Catalog # METBME104TE/00
Supplier:  Mettler - Toledo
Frequently Bought Together