MULTIWAVE GO PLUS 50Hz The Multiwave GO Plus with its newly developed Directed Multimode Cavity (DMC) offers the best of single-mode and multimode Microwave technology. This allows up to 12 samples at the same time open-minded and in combination with 1 * 1 ST
Catalog # PAAR130000
Supplier: Anton Paar
Frequently Bought Together