Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid protein, His Tag NUN-C51H9 is expressed from E.coli cells. It contains AA Met 1 - Ala 419 Accession QHO6211 5.1. 1 * 1 mg
ACRBNUN-C51H9-1MG
:
ACRO Biosystems
SARS-CoV-2 Nucleocapsid protein, His Tag NUN-C51H9 is expressed from E.coli cells. It contains AA Met 1 - Ala 419 Accession QHO6211 5.1. 1 * 1 mg
ACRBNUN-C51H9-1MG
:
ACRO Biosystems
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK