Mouse FCGRTB2M Heterodimer Protein, His TagTwin-Strep Tag BLI verified FCM-M52W2 is expressed from human 293 cells HEK293. It contai ns AA Ser 22 - Ser 297 FCGRT Ile 21 - Met 119 B2M Accession Q61559-1 FCGRT P01887-1 B2M. 1 * 50 µG
Catalog # ACRBFCM-M52W2-50UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together