ContactUsLinkComponent
SARS S1 protein, His Tag S1N-S52H5 is expressed from human 293 cells HEK293. It contains AA Ser 14 - Arg 667 Accession AAP13567.1. 1 * 100 µG
Catalog # ACRBS1N-S52H5100UG
Supplier:  ACRO Biosystems
SARS S1 protein, His Tag S1N-S52H5 is expressed from human 293 cells HEK293. It contains AA Ser 14 - Arg 667 Accession AAP13567.1. 1 * 100 µG
Catalog # ACRBS1N-S52H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together