ContactUsLinkComponent
Biotinylated Human CD3ECD3D Heterodimer Protein, His,AvitagTag Free CDD-H82W6 is expressed from human 293 cells HEK293. It contains AA Asp 23 - Asp 126 CD3E Phe 22 - Ala 105 CD3D Accession P07766-1 CD3E P04234-1 CD3D. 1 * 200 µG
Catalog # ACRBCDD-H82W6200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human CD3ECD3D Heterodimer Protein, His,AvitagTag Free CDD-H82W6 is expressed from human 293 cells HEK293. It contains AA Asp 23 - Asp 126 CD3E Phe 22 - Ala 105 CD3D Accession P07766-1 CD3E P04234-1 CD3D. 1 * 200 µG
Catalog # ACRBCDD-H82W6200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together