Human ILDR2, Fc Tag IL2-H5259 is expressed from human 293 cells HEK293. It contains AA Leu 21 - Glu 186 Accession Q71H61-1. 1 * 1 mg
ACRBIL2-H5259-1MG
Human ILDR2, Fc Tag IL2-H5259 is expressed from human 293 cells HEK293. It contains AA Leu 21 - Glu 186 Accession Q71H61-1. 1 * 1 mg
ACRBIL2-H5259-1MG
Frequently Bought Together