ContactUsLinkComponent
Biotinylated Human Transferrin R, His,Avitag TFR-H82E5 is expressed from human 293 cells HEK293. It contains AA Cys 89 - Phe 760 Acc ession P02786-1. 1 * 200 µG
Catalog # ACRBTFR-H82E5200UG
Supplier:  ACRO Biosystems
Biotinylated Human Transferrin R, His,Avitag TFR-H82E5 is expressed from human 293 cells HEK293. It contains AA Cys 89 - Phe 760 Acc ession P02786-1. 1 * 200 µG
Catalog # ACRBTFR-H82E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together