ContactUsLinkComponent
Human IL-17 RC, Fc Tag ILC-H5259 is expressed from human 293 cells HEK293. It contains AA Leu 21 - Ala 465 Accession NP_703190.2. 1 * 100 µG
Catalog # ACRBILC-H5259100UG
Supplier:  ACROBIOSYSTEMS
Human IL-17 RC, Fc Tag ILC-H5259 is expressed from human 293 cells HEK293. It contains AA Leu 21 - Ala 465 Accession NP_703190.2. 1 * 100 µG
Catalog # ACRBILC-H5259100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together