ContactUsLinkComponent
Biotinylated Human NKG2ACD94, Fc,Avitag™ NC4-H82F5 is expressed from human 293 cells HEK293. It contains AA Ala 113 - Leu 233 Asp 5 7-Ile 179 Accession P26715-1 NKG2A Q13241-1 CD94. 1 * 25 µG
Catalog # ACRBNC4-H82F5-25UG
Supplier:  ACRO Biosystems
Biotinylated Human NKG2ACD94, Fc,Avitag™ NC4-H82F5 is expressed from human 293 cells HEK293. It contains AA Ala 113 - Leu 233 Asp 5 7-Ile 179 Accession P26715-1 NKG2A Q13241-1 CD94. 1 * 25 µG
Catalog # ACRBNC4-H82F5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together