ContactUsLinkComponent
FITC-Labeled Human CD38, Fc Tag Cat. No. CD8-HF255 is expressed from human HEK293 cells. It contains AA Val 43 - Ile 300 Accession P28907-1. It is the FITC labeled form of Human CD38 Protein, Fc Tag Cat. No. CD8-H5255. 1 * 25 µG
Catalog # ACRBCD8-HF255-25UG
Supplier:  ACRO Biosystems
FITC-Labeled Human CD38, Fc Tag Cat. No. CD8-HF255 is expressed from human HEK293 cells. It contains AA Val 43 - Ile 300 Accession P28907-1. It is the FITC labeled form of Human CD38 Protein, Fc Tag Cat. No. CD8-H5255. 1 * 25 µG
Catalog # ACRBCD8-HF255-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together