FITC-Labeled Human CD27 Ligand, His,Flag Tag CDD-HF2D4 is expressed from human 293 cells HEK293. It contains AA Ser 52 - Pro 193 Acc ession P32970-1. It is the biotinylated form of Human CD27 Ligand, His,Flag Tag active trimer MALS verified Cat. No. 1 * 25 µG
ACRBCDD-HF2D4-25UG
Frequently Bought Together