ContactUsLinkComponent
MABSol® Biotinylated Mouse CD3ECD3D Heterodimer Protein, His,AvitagTag Free CDD-M82W5 is expressed from human HEK293 cells. It conta ins AA Asp 23 - Asp 108 CD3E Phe 22 - Ala 105 CD3D Accession P22646-1 CD3E P04235-1 CD3D. It is the biotinylated f 3 1 * 25 µG
Catalog # ACRBCDD-M82W5-25UG
Supplier:  ACROBIOSYSTEMS
MABSol® Biotinylated Mouse CD3ECD3D Heterodimer Protein, His,AvitagTag Free CDD-M82W5 is expressed from human HEK293 cells. It conta ins AA Asp 23 - Asp 108 CD3E Phe 22 - Ala 105 CD3D Accession P22646-1 CD3E P04235-1 CD3D. It is the biotinylated f 3 1 * 25 µG
Catalog # ACRBCDD-M82W5-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together