ContactUsLinkComponent
HCoV-OC43 Nucleocapsid, His Tag NUN-V5145 is expressed from E.coli cells. It contains AA Met 1 - Ile 448 Accession P33469-1. 1 * 100 µG
Catalog # ACRBNUN-V5145100UG
Supplier:  ACRO Biosystems
HCoV-OC43 Nucleocapsid, His Tag NUN-V5145 is expressed from E.coli cells. It contains AA Met 1 - Ile 448 Accession P33469-1. 1 * 100 µG
Catalog # ACRBNUN-V5145100UG
Supplier:  ACRO Biosystems
Frequently Bought Together