ContactUsLinkComponent
Biotinylated Cynomolgus FAP, His,Avitag™ FAP-C82Q5 is expressed from human 293 cells HEK293. It contains AA Leu 26 - Asp 760 Accessi on XP_005573377. 1 * 25 µG
Catalog # ACRBFAP-C82Q5-25UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus FAP, His,Avitag™ FAP-C82Q5 is expressed from human 293 cells HEK293. It contains AA Leu 26 - Asp 760 Accessi on XP_005573377. 1 * 25 µG
Catalog # ACRBFAP-C82Q5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together