ContactUsLinkComponent
MABSol® Biotinylated Human CD3ECD3D Heterodimer Protein, Fc,His,AvitagFc,Flag,Avitag CDD-H82F6 is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 CD3E Phe 22 - Ala 105 CD3D Accession P07766-1 CD3E P04234-1 CD3D. It is the bioti Hu 1 * 25 µG
Catalog # ACRBCDD-H82F6-25UG
Supplier:  ACRO Biosystems
MABSol® Biotinylated Human CD3ECD3D Heterodimer Protein, Fc,His,AvitagFc,Flag,Avitag CDD-H82F6 is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 CD3E Phe 22 - Ala 105 CD3D Accession P07766-1 CD3E P04234-1 CD3D. It is the bioti Hu 1 * 25 µG
Catalog # ACRBCDD-H82F6-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together