ContactUsLinkComponent
Biotinylated Mouse TGF-beta RII, His,Avitag™ TG2-M82H5 is expressed from human 293 cells HEK293. It contains AA Ile 24 - Asp 184 Acc ession Q62312-1. 1 * 25 µG
Catalog # ACRBTG2-M82H5-25UG
Supplier:  ACRO Biosystems
Biotinylated Mouse TGF-beta RII, His,Avitag™ TG2-M82H5 is expressed from human 293 cells HEK293. It contains AA Ile 24 - Asp 184 Acc ession Q62312-1. 1 * 25 µG
Catalog # ACRBTG2-M82H5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together