ContactUsLinkComponent
Biotinylated Human SIRP alpha V8, His,Avitag SI8-H82W6 is expressed from human 293 cells HEK293. It contains AA Glu 31- Arg 369 S44L , S50T, I52T, H54R, V57A Accession ATD50864.1. 1 * 200 µG
Catalog # ACRBSI8-H82W6200UG
Supplier:  ACRO Biosystems
Biotinylated Human SIRP alpha V8, His,Avitag SI8-H82W6 is expressed from human 293 cells HEK293. It contains AA Glu 31- Arg 369 S44L , S50T, I52T, H54R, V57A Accession ATD50864.1. 1 * 200 µG
Catalog # ACRBSI8-H82W6200UG
Supplier:  ACRO Biosystems
Frequently Bought Together