ContactUsLinkComponent
Biotinylated Human SIRP alphaV2, His,Avitag SI2-H82W8 is expressed from human 293 cells HEK293. It contains AA Glu 31 - Arg 369 Acce ssion ATD50864.1. 1 * 25 µG
  ACRBSI2-H82W8-25UG
Biotinylated Human SIRP alphaV2, His,Avitag SI2-H82W8 is expressed from human 293 cells HEK293. It contains AA Glu 31 - Arg 369 Acce ssion ATD50864.1. 1 * 25 µG
  ACRBSI2-H82W8-25UG
Frequently Bought Together