ContactUsLinkComponent
SARS-CoV-2 S protein, His Tag, Super stable trimer SPN-C52H7 is the ectodomain of SARS-CoV-2 S protein which contains AA Val 16 - Pr o 1213 Accession  QHD43416.1 R683A, R685A. The recombinant protein is expressed from human 293 cells HEK293 with T4 f 1 * 1 mg
Catalog # ACRBSPN-C52H7-1MG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 S protein, His Tag, Super stable trimer SPN-C52H7 is the ectodomain of SARS-CoV-2 S protein which contains AA Val 16 - Pr o 1213 Accession  QHD43416.1 R683A, R685A. The recombinant protein is expressed from human 293 cells HEK293 with T4 f 1 * 1 mg
Catalog # ACRBSPN-C52H7-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together