Human Kremen-1, Fc Tag KR1-H5253 is expressed from human 293 cells HEK293. It contains AA Arg 21 - Thr 392 Accession Q96MU8-1. 1 * 100 µG
ACRBKR1-H5253100UG
Human Kremen-1, Fc Tag KR1-H5253 is expressed from human 293 cells HEK293. It contains AA Arg 21 - Thr 392 Accession Q96MU8-1. 1 * 100 µG
ACRBKR1-H5253100UG
Frequently Bought Together