Rat CD3ECD3D Heterodimer Protein, Llama Fc TagLlama Fc Tag CDD-R5257 is expressed from human 293 cells HEK293. It contains AA Gln 21 - Asp 103 CD3E Phe 22 - Gly 106 CD3D Accession D4A5M2-1 CD3E P19377-1 CD3D. 1 * 1 mg
Rat CD3ECD3D Heterodimer Protein, Llama Fc TagLlama Fc Tag CDD-R5257 is expressed from human 293 cells HEK293. It contains AA Gln 21 - Asp 103 CD3E Phe 22 - Gly 106 CD3D Accession D4A5M2-1 CD3E P19377-1 CD3D. 1 * 1 mg