Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Cynomolgus TREM2, His Tag TR2-C52H3 is expressed from human 293 cells HEK293. It contains AA His 19 - Ser 174 Accession A0A2K5UY39- 1. 1 * 100 µG
ACRBTR2-C52H3100UG
:
ACRO Biosystems
Cynomolgus TREM2, His Tag TR2-C52H3 is expressed from human 293 cells HEK293. It contains AA His 19 - Ser 174 Accession A0A2K5UY39- 1. 1 * 100 µG
ACRBTR2-C52H3100UG
:
ACRO Biosystems
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Serological pipettes, Stripette™
: Corning
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Multiple well cell culture plates, Costar®
: Corning
VWR® Q Path®, Containers with Cap for Biopsy Probes
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
: EPPENDORF
VWR® PCR Tubes, 0,2 ml
: VWR Collection
Mouse Neuropilin-1 ELISA Kit
: Abcam
Human TREM2, His, Tag TR2-H52H5 is expressed from human 293 cells HEK293. It contains AA His 19 - Ser 174 Accession Q9NZC2-1. 1 * 100 µG
: ACRO Biosystems