ContactUsLinkComponent
Human CD30, His Tag CD0-H52H5 is expressed from human 293 cells HEK293. It contains AA Phe 19 - Lys 379 Accession NP_001234.2. 1 * 1 mg
Catalog # ACRBCD0-H52H5-1MG
Supplier:  ACRO Biosystems
Human CD30, His Tag CD0-H52H5 is expressed from human 293 cells HEK293. It contains AA Phe 19 - Lys 379 Accession NP_001234.2. 1 * 1 mg
Catalog # ACRBCD0-H52H5-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together