ContactUsLinkComponent
SARS-CoV-2 S protein RBD N501Y, His Tag SPD-C52Hn is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accessi on QHD43416.1 N501Y. N501Y mutaion was identified in both the SARS-CoV-2 variant known as 20B501Y.V1, VOC 20201201, a 1 * 100 µG
  ACRBSPD-C52HN100UG
SARS-CoV-2 S protein RBD N501Y, His Tag SPD-C52Hn is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accessi on QHD43416.1 N501Y. N501Y mutaion was identified in both the SARS-CoV-2 variant known as 20B501Y.V1, VOC 20201201, a 1 * 100 µG
  ACRBSPD-C52HN100UG
Frequently Bought Together