ContactUsLinkComponent
Human CD79ACD79B Heterodimer Protein, Mouse IgG2a Fc,Flag TagMouse IgG2a Fc,His Tag CDB-H52W3 is expressed from human 293 cells HEK2 93. It contains AA Leu 33 - Arg 143 CD79A Ala 29 - Asp 159 CD79B Accession P11912-1 CD79A P40259-1 CD79B. 1 * 1 mg
Catalog # ACRBCDB-H52W3-1MG
Supplier:  ACROBIOSYSTEMS
Human CD79ACD79B Heterodimer Protein, Mouse IgG2a Fc,Flag TagMouse IgG2a Fc,His Tag CDB-H52W3 is expressed from human 293 cells HEK2 93. It contains AA Leu 33 - Arg 143 CD79A Ala 29 - Asp 159 CD79B Accession P11912-1 CD79A P40259-1 CD79B. 1 * 1 mg
Catalog # ACRBCDB-H52W3-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together