ContactUsLinkComponent
Centrifuge tubes, 100 ml, round bottom, graduated, heavy wall, with screw cap, o.d. 44 mm, length 98 mm, nominal volume 85 ml 1 * 1 items
Catalog # WITG4.945.404
Supplier:  witeg Labortechnik
Centrifuge tubes, 100 ml, round bottom, graduated, heavy wall, with screw cap, o.d. 44 mm, length 98 mm, nominal volume 85 ml 1 * 1 items
Catalog # WITG4.945.404
Supplier:  witeg Labortechnik
Frequently Bought Together