Orders LinkContactUsLinkComponent
Core samplers
Core samplers
Core sampler, 800 mm, 25 mm, 230 ml
Catalog # 331-0376
Supplier:   BURKLE
Core samplers
Catalog # 331-0376
Supplier:  BURKLE
Core samplers
Core sampler, 800 mm, 25 mm, 230 ml
Price
642,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These core samplers can be used to take samples of bulk materials, which can easily become stuck or settle during sampling due to their particle form.
  • Stainless steel AISI 316
  • For bulk materials that become stuck or settle easily
In comparison to standard all-layer samplers for bulk materials, the two-part core sampler has no gap dimensions in which the medium can become stuck while it is being removed. This makes it possible to take a proper sample of powders and granulates through multiple layers, such as glass balls or metal powders.
Depending on the size, the stainless steel sampler can be used for a wide range of different containers, for example big bags or small containers. The bulk material sampler takes a representative core sample and is ideal for use in various laboratory and industrial applications.
 : Delivery includes core sampler (consisting of boring rod and boring tube) and fixing clip.
Specifications
  • Ø:
    25 mm
  • Volume:
    230 ml
  • Description:
    Core sampler
  • Length:
    800 mm
  • Seq:
    03