Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
EN 71-12:2013 Nitrosamines Mixture 121 1000 µg/mL in Methanol 1 * 1 mL
Catalog # EHERA50000121ME
Supplier:
EHRENSTORFER
Expand image
EN 71-12:2013 Nitrosamines Mixture 121 1000 µg/mL in Methanol 1 * 1 mL
Catalog # EHERA50000121ME
Supplier:
EHRENSTORFER
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals