HepaFilter H13 457x610x292mm HEPA Filter H13 (P-Line) , 457x610x292mm MDF-Rahmen, Mikro-Glasfaserpapier Filterklasse: H13 (EN1822) 250 Pa, 1750 m /h 0,9 m/s PU-Endlosdichtung einseitig 1 * 1 ST
Catalog # IBSAHM13PG457610
Supplier: IBS Stock Items
Frequently Bought Together