ContactUsLinkComponent
TM 50, temperature sensor, diam. 1,9 mm 0 to 120 °C for HD 3100/3200/3400/4050/4100/4200 for measuring the sample temperature 1 * 1 items
Catalog # BAND3733
TM 50, temperature sensor, diam. 1,9 mm 0 to 120 °C for HD 3100/3200/3400/4050/4100/4200 for measuring the sample temperature 1 * 1 items
Catalog # BAND3733
Frequently Bought Together