CHROMABOND lid (PP) SPE with gasket, for 24 columns (inclusive 24 Luer fittings for lid and 24 valves (plastic)) suitable for REF 7 30151N 1 * 24 items
CHROMABOND lid (PP) SPE with gasket, for 24 columns (inclusive 24 Luer fittings for lid and 24 valves (plastic)) suitable for REF 7 30151N 1 * 24 items