Combipack consisting of 702713: 1.5 mL Snap Ring Vial N 11 outer diameter: 11.6 mm, outer height: 32 mm clear, flat bottom, wide op ening, label + scale and 702710: N 11 PE snap ring cap, transparent Silicone white/PTFE red Hardness: 45° shore A Thic 1 * 1.000 items
Catalog # MANA702721
Supplier: MACHEREY-NAGEL
Frequently Bought Together